missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAD3 Recombinant Rabbit Monoclonal Antibody (8M7B3)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA538011
This item is not returnable.
View return policy
Description
Positive Samples: A-549, DU145, Jurkat Immunogen sequence: RRARMHIQKL EDQEQRARQL KERLRSKQQS LQRQLEQLRG LAGAAERERL RADSLDSSGL SSERSDSDQE ELEVDVESLV FGGEAELLRG F.
This gene encodes a member of the Myc superfamily of basic helix-loop-helix leucine zipper transcriptional regulators. The encoded protein forms a heterodimer with the cofactor MAX which binds specific E-box DNA motifs in the promoters of target genes and regulates their transcription. Disruption of the MAX-MXD3 complex is associated with uncontrolled cell proliferation and tumorigenesis. Transcript variants of this gene encoding different isoforms have been described.
Specifications
| MAD3 | |
| Recombinant Monoclonal | |
| 0.46 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9BW11 | |
| MXD3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 100-190 of human MAD3/MXD3 (Q9BW11). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 8M7B3 | |
| Unconjugated | |
| MXD3 | |
| 4631412E13Rik; bHLHc13; class C basic helix-loop-helix protein 13; FLJ35523; MAD3; max dimerization protein 3; Max dimerizer 3; Max-associated protein 3; Max-interacting transcriptional repressor MAD3; MGC2383; Mxd3; Myx | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 83463 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction