missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAD3 Rabbit anti-Human, Mouse, Rat, Clone: 8M7B3, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | MAD3 |
|---|---|
| Clone | 8M7B3 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18346163
|
Bio-Techne
NBP3-15929-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18344416
|
Novus Biologicals
NBP3-15929-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MAD3 Monoclonal antibody specifically detects MAD3 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| MAD3 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 83463 | |
| IgG | |
| Affinity purified |
| 8M7B3 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| bHLHc13, Class C basic helix-loop-helix protein 13, MAD3FLJ35523, MAX dimerization protein 3, Max dimerizer 3, Max-associated protein 3, Max-interacting transcriptional repressor MAD3, MGC2383, MYX | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 100-190 of human MAD3 (Q9BW11). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto