missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAD3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568499
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 93%; Dog: 86%; Horse: 93%; Human: 100%; Mouse: 85%; Pig: 86%; Rabbit: 86%; Rat: 93%.
This gene encodes a member of the Myc superfamily of basic helix-loop-helix leucine zipper transcriptional regulators. The encoded protein forms a heterodimer with the cofactor MAX which binds specific E-box DNA motifs in the promoters of target genes and regulates their transcription. Disruption of the MAX-MXD3 complex is associated with uncontrolled cell proliferation and tumorigenesis. Transcript variants of this gene encoding different isoforms have been described.
Specifications
| MAD3 | |
| Polyclonal | |
| Unconjugated | |
| MXD3 | |
| 4631412E13Rik; bHLHc13; class C basic helix-loop-helix protein 13; FLJ35523; MAD3; max dimerization protein 3; Max dimerizer 3; Max-associated protein 3; Max-interacting transcriptional repressor MAD3; MGC2383; Mxd3; Myx | |
| Rabbit | |
| Protein A | |
| RUO | |
| 83463 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9BW11 | |
| MXD3 | |
| synthetic peptide directed towards the N terminal region of human MAD3. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction