missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAD2L2 Recombinant Rabbit Monoclonal Antibody (24GB4950)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557482
This item is not returnable.
View return policy
Description
MAD2L2 Recombinant Monoclonal Antibody for Western Blot, Flow
The protein encoded by this gene is a component of the mitotic spindle assembly checkpoint that prevents the onset of anaphase until all chromosomes are properly aligned at the metaphase plate. The encoded protein, which is similar to MAD2L1, is capable of interacting with ADAM9, ADAM15, REV1, and REV3 proteins.
Specifications
| MAD2L2 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| MAD2L2 | |
| 2310033C13Rik; G1-453-4; hREV7; MAD2 (mitotic arrest deficient, yeast, homolog)-like 2; MAD2 mitotic arrest deficient-like 2; MAD2 mitotic arrest deficient-like 2 (yeast); Mad2b; Mad2l2; MAD2-like protein 2; Mitotic arrest deficient 2-like protein 2; mitotic arrest deficient homolog-like 2; mitotic spindle assembly checkpoint protein MAD2B; polymerase (DNA-directed), zeta 2, accessory subunit; POLZ2; repro22; REV7; REV7 homolog; RP3-330O12.4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10459 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB4950 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9UI95 | |
| MAD2L2 | |
| A synthesized peptide derived from human Mad2L2 (aa 178-191). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction