missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAD2L2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38622-20ul
This item is not returnable.
View return policy
Description
MAD2L2 Polyclonal antibody specifically detects MAD2L2 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| MAD2L2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| hREV7, MAD2 mitotic arrest deficient-like 2 (yeast), MAD2BREV7 homolog, MAD2-like protein 2, Mitotic arrest deficient 2-like protein 2, mitotic arrest deficient homolog-like 2, mitotic spindle assembly checkpoint protein MAD2B, REV7yeast, homolog)-like 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-211 of human MAD2L2 (NP_006332.3).,, Sequence:, MTTLTRQDLNFGQVVADVLCEFLEVAVHLILYVREVYPVGIFQKRKKYNVPVQMSCHPELNQYIQDTLHCVKPLLEKNDVEKVVVVILDKEHRPVEKFVFEITQPPLLSISSDSLLSHVEQLLRAFILKISVCDAVLDHNPPGCTFTVLVHTREAATRNMEKIQVIKDFPWILADEQDVHMHDPRLIPLKTMTSDILKMQLYVEERAHKGS | |
| 20 μL | |
| Cell Cycle and Replication, Phospho Specific | |
| 10459 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction