missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAD2L1BP, Mouse anti-Human, Clone: 4G11, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00009587-M03.100ug
This item is not returnable.
View return policy
Description
Sequence: MAAPEAEVLSSAAVPDLEWYEKSEETHASQIELLETSSTQEPLNASEAFCPRDCMVPVVFPGPVSQEGCCQFTCELLKHIMYQRQQLPLPYEQLKHFYRKPSPQAEEMLKKKPRATTEVSSRKCQQALAELESVLSHLEDFFARTLVPRVLILLGGNALSPKEFYELDLSLLAPYSVDQSLSTAACLRRLFRAIFMADAFSELQAPPLMGTVVMAQGHRNCGEDWFRPKLNYRVPSRGHKLTVTLSCGRPSIRTTAWEDYIWFQAPVTFKGFRESpecifications
| MAD2L1BP | |
| Monoclonal | |
| Unconjugated | |
| MAD2L1 binding protein | |
| CMT2/KIAA0110/MGC11282/RP1-261G23.6 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Western Blot | |
| 4G11 | |
| In 1x PBS, pH 7.4 | |
| BC002904 | |
| MAD2L1BP | |
| MAD2L1BP (AAH02904, 1 a.a. ∼ 274 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 9587 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction