missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAD2L1BP Monoclonal Antibody (OTI3C11)

MAD2L1BP Monoclonal Antibody (OTI3C11)
Brand: Invitrogen MA526699
This item is not returnable.
View return policy
Description
Specifications
| MAD2L1BP | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q15013 | |
| MAD2L1BP | |
| Full length human recombinant protein of MAD2L1BP produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3C11 | |
| Unconjugated | |
| MAD2L1BP | |
| 0610009D16Rik; 2510031P20Rik; AU018945; AW550977; C76045; caught by MAD2 protein; CMT2; CMT2A; CMT2B; KIAA0110; MAD2L1 binding protein; MAD2L1-binding protein; Mad2l1bp; Mad2lbp; mKIAA0110; p31(comet); RP1-261G23.6 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9587 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction