missing translation for 'onlineSavingsMsg'
Learn More

MAD2L1 Antibody (OTI4D2), Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18411387 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18411387 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18411387 Supplier Novus Biologicals Supplier No. NBP272565AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MAD2L1 Monoclonal antibody specifically detects MAD2L1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAD2L1
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone OTI4D2
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias HSMAD2, MAD2 (mitotic arrest deficient, yeast, homolog)-like 1, MAD2 mitotic arrest deficient-like 1 (yeast), MAD2-like protein 1, MAD2mitotic arrest deficient, yeast, homolog-like 1, Mitotic arrest deficient 2-like protein 1, mitotic spindle assembly checkpoint protein MAD2A
Host Species Mouse
Immunogen Full length recombinant protein of human MAD2L1 (NP_002349) produced in HEK293T cells.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4085
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.