missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAD2 Polyclonal Antibody

MAD2 Polyclonal Antibody
Brand: Invitrogen PA586716
This item is not returnable.
View return policy
Description
MAD2 Polyclonal Antibody for Western Blot
Specifications
| MAD2 | |
| Polyclonal | |
| Unconjugated | |
| MAD2L1 | |
| AA673185; HsMAD2; J1256; MAD2; MAD2 (mitotic arrest deficient, homolog)-like 1; MAD2 (mitotic arrest deficient, yeast, homolog)-like 1; MAD2 mitotic arrest deficient-like 1; MAD2 mitotic arrest deficient-like 1 (yeast); Mad2a; MAD2L1; MAD2-like 1; MAD2-like protein 1; MadL1; Mitotic arrest deficient 2-like protein 1; mitotic arrest deficient, yeast, homolog-like 1; Mitotic MAD2 protein; Mitotic spindle assembly checkpoint protein MAD2A; Mitotic spindle checkpoint component MAD2; spindle checkpoint protein MAD2; YJL030W | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 297176, 4085, 56150 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q13257, Q9Z1B5 | |
| MAD2L1 | |
| Synthetic peptide, corresponding to amino acids 134-179 of Human MAD2L1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction