missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAD1L1/MAD1 Rabbit anti-Human, Mouse, Clone: 7O2C2, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | MAD1L1/MAD1 |
|---|---|
| Clone | 7O2C2 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18336564
|
Novus Biologicals
NBP3-16553-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18345052
|
Bio-Techne
NBP3-16553-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MAD1L1/MAD1 Monoclonal antibody specifically detects MAD1L1/MAD1 in Human, Mouse samples. It is validated for Western BlotSpecifications
| MAD1L1/MAD1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Cycle and Replication, DNA Repair, Tumor Suppressors | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 8379 | |
| IgG | |
| Affinity purified |
| 7O2C2 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| hMAD1, MAD1 mitotic arrest deficient-like 1 (yeast), MAD1-like protein 1, MAD1yeast, homolog-like 1, Mitotic checkpoint MAD1 protein homolog, Tax-binding protein 181, tumor protein p53 inducible protein 9, TXBP181MAD1 (mitotic arrest deficient, yeast, homolog)-like 1 | |
| A synthetic peptide corresponding to a sequence within amino acids 50-150 of human MAD1L1/MAD1 (Q9Y6D9). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title