missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAD1L1/MAD1 Antibody (JE35-54), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32556
This item is not returnable.
View return policy
Description
MAD1L1/MAD1 Monoclonal antibody specifically detects MAD1L1/MAD1 in Human samples. It is validated for Western Blot
Specifications
| MAD1L1/MAD1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000 | |
| hMAD1, MAD1 mitotic arrest deficient-like 1 (yeast), MAD1-like protein 1, MAD1yeast, homolog-like 1, Mitotic checkpoint MAD1 protein homolog, Tax-binding protein 181, tumor protein p53 inducible protein 9, TXBP181MAD1 (mitotic arrest deficient, yeast, homolog)-like 1 | |
| Recombinant protein within Human MAD1L1/MAD1 aa 31-180/ 718. (Uniprot: Q9Y6D9) | |
| 100 μL | |
| Cancer, Cell Cycle and Replication, DNA Repair, Tumor Suppressors | |
| 8379 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| JE35-54 | |
| Unconjugated | |
| 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction