missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAD1L1/MAD1 Antibody (9B10), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-5783JF549
This item is not returnable.
View return policy
Description
MAD1L1/MAD1 Monoclonal specifically detects MAD1L1/MAD1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| MAD1L1/MAD1 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MAD1L1 | |
| Recombinant full length human MAD1 protein. | |
| 0.1 mL | |
| Primary | |
| MAD1 (9B10) | |
| Store at 4°C in the dark. | |
| IgG2b |
| Immunocytochemistry, Immunofluorescence | |
| 9B10 | |
| Immunocytochemistry/Immunofluorescence | |
| hMAD1, MAD1 mitotic arrest deficient-like 1 (yeast), MAD1-like protein 1, MAD1yeast, homolog-like 1, Mitotic checkpoint MAD1 protein homolog, Tax-binding protein 181, tumor protein p53 inducible protein 9, TXBP181MAD1 (mitotic arrest deficient, yeast, homolog)-like 1 | |
| Mouse | |
| Protein A or G purified | |
| Cancer, Cell Cycle and Replication, DNA Repair, Tumor Suppressors | |
| 8379 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction