missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MAD1 Recombinant Rabbit Monoclonal Antibody (24GB3620)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£126.00 - £434.00

Specifications

Antigen MAD1
Clone 24GB3620
Content And Storage -20°C
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30275053
GREENER_CHOICE
View Documents
Invitrogen™
MA557079
100 μL
£434.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30276037
View Documents
Invitrogen™
MA557080
20 μL
£126.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MAD1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

MAX dimerization protein belongs to a subfamily of MAX-interacting proteins. This protein competes with MYC for binding to MAX to form a sequence-specific DNA-binding complex, acts as a transcriptional repressor (while MYC appears to function as an activator) and is a candidate tumor suppressor.
Specifications

Specifications

MAD1
-20°C
Recombinant Monoclonal
Liquid
IgG
Human, Mouse, Rat
MAD1L1
AI173502; AW550425; coiled-coil domain-containing protein MAD1; hMAD1; HsMAD1; Mad1; MAD1 mitotic arrest deficient 1-like 1; MAD1 mitotic arrest deficient like 1; MAD1 mitotic arrest deficient-like 1; MAD1 mitotic arrest deficient-like 1 (yeast); MAD1L1; MAD1-like protein 1; mitotic arrest deficient 1-like 1; mitotic arrest deficient 1-like protein 1; Mitotic arrest deficient protein 1; Mitotic checkpoint MAD1 protein homolog; mitotic spindle assembly checkpoint protein MAD1; mitotic-arrest deficient 1, yeast, homolog-like 1; MXD1; PIG9; Spindle assembly checkpoint component MAD1; Tax-binding protein 181; TP53I9; tumor protein p53 inducible protein 9; TXBP181; YGL086W
MAD1L1
Primary
Affinity chromatography
24GB3620
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q9WTX8, Q9Y6D9
17120, 680006, 8379
A synthesized peptide derived from human MAD1 (aa 50-170).
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.