Learn More
Cytiva Macrosep™ Advance Centrifugal Devices with Omega Membrane
Product Code. 17154731
Change view
Click to view available options
MWCO:
1 kDa
10 kDa
100 kDa
3 kDa
30 kDa
Includes:
100/Pk.
24/Pk.
Color:
Blue
Clear
Gray
Red
Yellow
Unit Size:
Pack of 100
Pack of 24
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. MWCO Includes Color unitSize
17154731 3 kDa 100/Pk. Gray Pack of 100
17174731 10 kDa 100/Pk. Blue Pack of 100
17124731 1 kDa 24/Pk. Yellow Pack of 24
17184731 30 kDa 24/Pk. Red Pack of 24
17104741 100 kDa 24/Pk. Clear Pack of 24
17134731 1 kDa 100/Pk. Yellow Pack of 100
17164731 10 kDa 24/Pk. Blue Pack of 24
17114741 100 kDa 100/Pk. Clear Pack of 100
17194731 30 kDa 100/Pk. Red Pack of 100
17144731 3 kDa 24/Pk. Gray Pack of 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 17154731 Supplier Cytiva Supplier No. MAP003C38
  • ONLINE EXCLUSIVE PRICE
    £1085.00
    £1024.00 / Pack of 100
    Save £61.00
    5% Off
View all promotions
In Stock
Add to basket
Add to basket
Macrosep Advance Centrifugal Devices with Omega Membrane quickly concentrates up to 20 mL of biological sample.
  • Exchange buffer or remove salt of chromatography eluates and gradient fractions
  • For research applications only
TRUSTED_SUSTAINABILITY
Specifications
Color Gray
Description EFA: 7.2 cm2
Material (Membrane) PES
MWCO 3 kDa
Sterility Non-sterile
Includes 100/Pk.
Volume (Metric) 5 to 20 mL (Sample)