Learn More
Macrophages/Monocytes Monoclonal Antibody (MOMA-2), PE, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA516865
Description
This clone recognizes an intracellular antigen of mouse macrophages and monocytes. It reacts strongly with macrophages in lymphoid organs such as tingible body macrophages and macrophages in T cell dependant areas. Membrane permeabilization is required for Flow Cytometry. Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4°C in the dark.
Macrophages are a large phagocytic cell found in stationary form in the tissues or as a mobile white blood cell, especially at sites of infection. Monocytes are a large phagocytic white blood cell with a simple oval nucleus and clear, grayish cytoplasm. Monocytes and macrophages are found in interdigitating cells and activated microglia cells.
Specifications
| Macrophages/Monocytes | |
| Monoclonal | |
| Conc. Not Determined | |
| PBS with 1% BSA, 5% sucrose and 0.09% sodium azide | |
| Rat | |
| Protein G | |
| RUO | |
| Mouse | |
| Antibody | |
| IgG2b |
| Flow Cytometry | |
| MOMA-2 | |
| PE | |
| Macrophage; Monocyte | |
| Mouse lymph node stroma | |
| 100 Tests | |
| Primary | |
| 4° C, store in dark, DO NOT FREEZE! | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.