missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Macrophages/Monocytes Monoclonal Antibody (KUL01), Biotin
GREENER_CHOICE

Product Code. 16313084
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16313084 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16313084 Supplier Invitrogen™ Supplier No. MA528826

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Macrophages/Monocytes Monoclonal Antibody for Flow

Macrophages are a large phagocytic cell found in stationary form in the tissues or as a mobile white blood cell, especially at sites of infection. Monocytes are a large phagocytic white blood cell with a simple oval nucleus and clear, grayish cytoplasm. Monocytes and macrophages are found in interdigitating cells and activated microglia cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Macrophages/Monocytes
Applications Flow Cytometry
Classification Monoclonal
Clone KUL01
Concentration 0.5 mg/mL
Conjugate Biotin
Formulation PBS with Proprietary stabilizer and <0.1% sodium azide; pH 7.4
Host Species Mouse
Immunogen Chicken peripheral blood mononuclear cell leukocytes.
Purification Method Purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chicken
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.