missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macrophage Marker Antibody (5/9), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50482F
This item is not returnable.
View return policy
Description
Macrophage Marker Monoclonal specifically detects Macrophage Marker in Human samples. It is validated for Immunohistochemistry.
Specifications
| Macrophage Marker | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| A cell suspension containing osteoclasts from osteoclastomas. | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. |
| Immunohistochemistry | |
| 5/9 | |
| Immunohistochemistry | |
| Mouse | |
| Protein A purified | |
| Immunology | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction