Learn More
Berthold Technologies Macro Table With Temp. Control
Product Code. 31777402 Shop All Berthold Technologies Products
Change view
Click to view available options
For Use With (Application):
Imaging Small Objects
Unit Size:
Each
Product Code. For Use With (Application) Unit Size
31777402 Imaging Small Objects Each
1 options
Product Code. 31777402 Supplier Berthold Technologies Supplier No. 51578
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Provides additional magnification for the Berthold NightSHADE and NightOWL imaging systems.
  • Reducing the distance between camera and sample increases photon collection due to lens solid angle
  • Suitable for imaging small objects
  • The macro table enables magnification up to 5X
  • Integrated temperature control supports imaging of temperature-sensitive samples, including living specimens
Specifications
Autoclavable Not Autoclavable
Animal Mouse
For Use With (Application) Imaging Small Objects
Disposable Reusable
Size One Size
Sterility Non-sterile
Voltage 24 V