missing translation for 'onlineSavingsMsg'
Learn More

Macranthoidin B, MedChemExpress

Product Code. 16449148
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16449148 10 mg 10mg
16401558 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16449148 Supplier Medchem Express Supplier No. HYN0864
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Macranthoidin B is a major bioactive saponin in rat plasma after oral administration of extraction of saponins from Flos Lonicerae.

Macranthoidin B is a major bioactive saponin in rat plasma after oral administration of extraction of saponins from Flos Lonicerae.

Specifications

Chemical Name or Material Macranthoidin B
CAS 136849-88-2
Color White
Purity Grade Notes Research
Molecular Formula C65H106O32
Quantity 10 mg
Synonym Macranthoiside I
Solubility Information H2O : 100 mg/mL (71.45 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (71.45 mM; Need ultrasonic)
SMILES O=C([C@]1(CCC(C)(C)C2)[C@]2([H])C3=CC[C@@]4([H])[C@@](C)(CC[C@]5([H])[C@@]4(CC[C@H](O[C@@](OC[C@H](O)[C@@H]6O)([H])[C@@H]6O[C@@](O[C@@H](C)[C@H](O)[C@H]7O[C@]([C@@H]([C@@H](O)[C@@H]8O[C@]([C@@H]([C@@H](O)[C@@H]9O)O)([H])O[C@@H]9CO)O)([H])O[C@@H]8CO)([H])[C@@H]7O)[C@@]5(C)CO)C)[C@]3(C)CC1)O[C@@H]([C@@H]([C@@H](O)[C@@H]%10O)O)O[C@@H]%10CO[C@@H]([C@@H]([C@@H](O)[C@@H]%11O)O)O[C@@H]%11CO
Molecular Weight (g/mol) 1399.52
Formula Weight 1399.52
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.