missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MACF1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13058-1-AP-150UL
This item is not returnable.
View return policy
Description
SNARE that acts to regulate protein transport between late endosomes and the trans-Golgi network. The SNARE complex containing STX6, STX12, VAMP4 and VTI1A mediates vesicle fusion (in vitro).Specifications
| MACF1 | |
| Polyclonal | |
| Unconjugated | |
| MACF1 | |
| 620 kDa actin binding protein, ABP620, ACF7, KIAA0465, KIAA1251, MACF, MACF1, Macrophin 1, OFC4, Trabeculin alpha | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11426, 23499, 362587 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| D3ZHV2, Q9QXZ0, Q9UPN3 | |
| Macf1 | |
| MACF1 Fusion Protein Ag3730 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction