missing translation for 'onlineSavingsMsg'
Learn More

Macelignan, MedChemExpress

Product Code. p-200100043
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16488848

Brand: Medchem Express HYN0064

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Macelignan ((+)-Anwulignan; Anwuligan) is an orally active lignan isolated from Myristica fragrans. Macelignan possesses many pharmacological activities, including anti-inflammatory, anti-cancer, anti-diabetes, and neuroprotective activities.

Macelignan ((+)-Anwulignan

Specifications

Chemical Name or Material Macelignan
CAS 107534-93-0
Color White
Purity Grade Notes Research
Molecular Formula C20H24O4
Quantity 10 mg
Synonym (+)-Anwulignan Anwuligan
Solubility Information DMSO : 100 mg/mL (304.51 mM; Need ultrasonic)
SMILES OC1=CC=C(C[C@H](C)[C@H](C)CC2=CC=C(OCO3)C3=C2)C=C1OC
Molecular Weight (g/mol) 328.4
Formula Weight 328.4
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.