missing translation for 'onlineSavingsMsg'
Learn More

Macelignan, MedChemExpress

Product Code. 16488848
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16488848 10 mg 10mg
16401298 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16488848 Supplier Medchem Express Supplier No. HYN0064
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Macelignan ((+)-Anwulignan; Anwuligan) is an orally active lignan isolated from Myristica fragrans. Macelignan possesses many pharmacological activities, including anti-inflammatory, anti-cancer, anti-diabetes, and neuroprotective activities.

Macelignan ((+)-Anwulignan

Specifications

Chemical Name or Material Macelignan
CAS 107534-93-0
Color White
Purity Grade Notes Research
Molecular Formula C20H24O4
Quantity 10 mg
Synonym (+)-Anwulignan Anwuligan
Solubility Information DMSO : 100 mg/mL (304.51 mM; Need ultrasonic)
SMILES OC1=CC=C(C[C@H](C)[C@H](C)CC2=CC=C(OCO3)C3=C2)C=C1OC
Molecular Weight (g/mol) 328.4
Formula Weight 328.4
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.