Learn More
Emma Safety Footwear Mace M Safety Shoes
A superb shoe for galvanizing and welding metal. Closed front flap protects against stray sparks and hot metal splinters. Hydro-Tec™ Sanitized Silver keep feet dry and fresh.
Brand:  Emma Safety Footwear MM50486337
 View more versions of this product
Product Code. 15988471
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • S3-Safety shoe with sole plate for wet work
  • GripForce™ Duo / HRO
  • Full grain leather
  • Heat resistant outsole
  • Hydro-Tec™ Sanitized Silver
Specifications
Specifications
EN ISO 20345:2011
Black
Hydro-Tec™ Sanitized Silver
37
Laces
Mace M
Leather
Steel