missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MacConkey Agar No. 3 (Dehydrated)
GREENER_CHOICE

Product Code. 10239762
Change view
Click to view available options
Quantity:
500 g
2.5kg
5 kg
Unit Size:
2.50kg
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10239762 5 kg 5kg
10229762 500 g 500g
15823207 2.5kg 2.50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10239762 Supplier Thermo Scientific™ Supplier No. CM0115T

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Detect and enumerate coliform organisms, including E. coli, while isolating and detecting Salmonella and Shigella spp. with this selective medium.

Detect and enumerate coliform organisms, including Escherichia coli, while isolating and detecting Salmonella and Shigella spp. in both clinical and food specimens using Thermo Scientific™ Oxoid™ MacConkey Agar No.3. A modification of the original MacConkey medium by the addition of selected bile salts, the medium gives improved differentiation of coliforms and non-lactose fermenting organisms while completely inhibiting Gram-positive cocci.

Use MacConkey Agar No. 3 for detection and enumeration of coliform organisms while isolating and detecting Salmonella and Shigella spp. in pathological and food specimens.

  • Selective and differential: due to the inclusion of a specially prepared fraction of bile salts in addition to crystal violet, the medium gives improved differentiation between coliforms and non-lactose fermenting organisms while Gram-positive cocci are completely inhibited.
  • Easy-to-read: Escherichia coli (red colonies with bile precipitation) is easy to differentiate from Shigella spp. (straw coloured colonies) and Salmonella (Straw coloured colonies).
  • Recommended by the American Public Health Association 1

MacConkey Agar No.3 formulation corresponds with that recommended by the American Public Health Association1 for the direct plating of water samples for coliform bacilli, for the examination of food samples for food poisoning organisms2 and for the isolation of Salmonella and Shigella species in cheese3.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Form Powder
Product Type Agar
Quantity 5 kg

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.