Learn More
Neogen Macadamia Protein ELISA Kit, E96MAC
Product Code. 17273814
Encompass
Click to view available options
Test Time:
50 Minutes
Unit Size:
1 set
Product Code. 17273814
Brand: Neogen 7100151368
Intended for the detection of macadamia proteins in clean-in-place water (CIP) final rinse water, environmental swab samples, food ingredients and processed food products.
  • Designed to detect both processed and unprocessed target allergen proteins.
  • Test can accommodate environmental swab, CIP rinse water and food samples.
  • A universal extraction protocol streamlines testing for multiple analytes, saving you reagents and valuable time.
Specifications
Product Type Macadamia Protein ELISA Kit
Test Time 50 Minutes