missing translation for 'onlineSavingsMsg'
Learn More

MAC13243, MedChemExpress

Product Code. p-200099409
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16478677

Brand: Medchem Express HY14456A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MAC13243, an antibacterial agent, is an inhibitor of bacterial lipoprotein targeting chaperone, LolA. MAC13243 is an antibacterial agent with Gram-negative selectivity.

MAC13243, an antibacterial agent, is an inhibitor of bacterial lipoprotein targeting chaperone, LolA. MAC13243 is an antibacterial agent with Gram-negative selectivity.

Specifications

Chemical Name or Material MAC13243
Physical Form Solid
CAS 1071638-38-4
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C20H25Cl2N3O2S
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 50 mg/mL (113.02 mM; Need ultrasonic)
SMILES ClC1=CC=C(CSC2=NCN(CCC3=CC(OC)=C(OC)C=C3)CN2)C=C1.Cl
Molecular Weight (g/mol) 442.4
Formula Weight 442.4
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.