missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MAbPac™ SEC-1 Size Exclusion Chromatography HPLC Columns

Product Code. 11392073
Change view
Click to view available options
Length (Metric):
50 mm
150 mm
300 mm
Diameter (Metric):
2.1 mm
4 mm
7.8 mm
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) unitSize
11392073 150 mm 4 mm Each
15320421 300 mm 7.8 mm Each
11302073 300 mm 4 mm Each
11312073 50 mm 4 mm Each
15590244 150 mm 2.1 mm Each
15580244 300 mm 2.1 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 11392073 Supplier Thermo Scientific™ Supplier No. 075592
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Use these SEC columns for monoclonal antibody (mAb) analysis, including the high-resolution separation of monomers, aggregates and fragments.

Use Thermo Scientific™ MAbPac™ SEC-1 Size Exclusion Chromatography HPLC Columns for high-resolution separation of monoclonal antibody (mAb) analysis, including the monomers, aggregates and Fab and Fc fragments resulting from proteolysis. These size-exclusion chromatography (SEC) columns provide superior performance, even under nondenaturing conditions, and in high- and low-salt mobile phases and volatile eluents.

High-Performance SEC Columns for MAb Analysis

  • Hydrophilic bonded layer minimizes undesired interactions between the biomolecules and the stationary phase
  • Nonmetallic and biocompatible PEEK housing eliminates metal contamination from the column hardware
  • Stable surface bonding leads to low column bleed and compatibility with MS, ELSD and Thermo Scientific™ Dionex™ Corona™ CAD detection
  • Rugged column packing provides reliability, reproducibility and long column life
  • Columns are available in 5μm particle size and 150 and 300mm lengths, with an average pore diameter of 300Å
TRUSTED_SUSTAINABILITY

Specifications

Column Chemistry Proprietary Diol
Column Format Analytical Column
Diameter (Metric) 4 mm
Length (Metric) 150 mm
Max. Pressure 600 psi (40 bar)
Particle Size 5 μm
pH 2.5 to 7.5
Pore Size 300 Å
Quantity Each
Solvents 100% Organic Solvents
Temperature 30°C
USP Type L59
Stationary Phase Diol (OH)
Product Line MAbPac
Packing Material Spherical, Fully Porous, Ultrapure Silica
Column Type Size Exclusion
Phase Size Exclusion
Material PEEK
Technique Bio-LC
Green Features FSC
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.