missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MAbPac™ SCX-10 HPLC Columns

Product Code. 11391993
Change view
Click to view available options
Length (Metric):
50 mm
150 mm
250 mm
Diameter (Metric):
2 mm
4 mm
4.6 mm
9 mm
Particle Size:
3 μm
5 μm
10 μm
Column Format:
Analytical Column
Guard Column
Preparative Column
Unit Size:
Each
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Particle Size Column Format unitSize
11391993 250 mm 2 mm 10 μm Analytical Column Each
11372063 250 mm 4 mm 10 μm Analytical Column Each
11332083 150 mm 4 mm 10 μm Analytical Column Each
11342083 50 mm 4 mm 10 μm Analytical Column Each
15540244 250 mm 9 mm 10 μm Preparative Column Each
15124725 150 mm 4 mm 5 μm Analytical Column Each
15134725 50 mm 4.6 mm 5 μm Analytical Column Each
15154725 150 mm 4.6 mm 5 μm Analytical Column Each
15144725 250 mm 4.6 mm 5 μm Analytical Column Each
12663085 50 mm 4 mm 3 μm Analytical Column Each
12683085 250 mm 4 mm 5 μm Analytical Column Each
12673085 50 mm 4 mm 5 μm Analytical Column Each
11382023 50 mm 2 mm 10 μm Guard Column Each
11362063 50 mm 4 mm 10 μm Guard Column Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
This item is not returnable. View return policy
Product Code. 11391993 Supplier Thermo Scientific™ Supplier No. 075604
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Use these strong cation-exchange columns for high-resolution, high efficiency analysis of monoclonal antibodies (mAbs) and associated variants.

Use Thermo Scientific™ MAbPac™ SCX-10 LC Columns for high-resolution, high efficiency analysis of monoclonal antibodies (mAbs) and associated variants. The unique nonporous pellicular resin provides the resolving power to separate mAb variants that differ by as little as one charged residue. A hydrophilic layer surrounds the polymeric beads and eliminates hydrophobic interactions with the resin, resulting in very efficient peaks. A proprietary grafted cation-exchange surface provides pH selectivity control for high-resolution separations.

Superior Performance for mAb Variant Analysis
  • Exceptionally high resolution for mAb variants
  • Ideal for characterization and quality control assessment of mAbs
  • Well-suited for stability studies, mAb therapeutic method development and high-throughput screening
  •  Unmatched column-to-column and lot-to-lot reproducibility eliminates column variability as a concern in stability and QA/QC testing
  • Meets the regulatory requirements for biopharmaceutical characterization
Multiple Sizes Available
  • Available in 3, 5 and 10μm particle sizes in standard and UHPLC formats
  • Smaller-particle-size columns provide the same resolution as the 10μm columns, but with significantly faster analysis time
TRUSTED_SUSTAINABILITY

Specifications

Column Format Analytical Column
Diameter (Metric) 2 mm
Flow Rate 0.25 mL/min
Length (Metric) 250 mm
Max. Pressure 3000 psi (207 bar)
Particle Size 10 μm
pH 2 to 12
Pore Size Nonporous
Quantity Each
Solvents 50% Acetonitrile
Temperature Ambient to 60°C
Stationary Phase Sulfonate
Product Line MAbPac
Packing Material Spherical, Nonporous Polymer
Column Type Ion Exchange
Phase Ion Exchange
Type Analytical Column
Technique Bio-LC
Green Features FSC
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.