Learn More
Thermo Scientific™ MAbPac™ HIC-10 HPLC Column
Separate monoclonal antibodies (mAbs) and related biologics with high resolution, excellent biocompatibility, and selectivity using the unique chemistry of this hydrophobic interaction column.
Brand: Thermo Scientific™ 088481
Code : NEW
Additional Details : Weight : 0.44000kg
Description
Designed for separation of monoclonal antibodies (mAbs) and related biologics by hydrophobic interaction, the Thermo Scientific™ MAbPac™ HIC-10 LC column is a high resolution silica column. The MAbPac HIC-10 column's unique chemistry provides high resolution, excellent biocompatibility, and selectivity that is different from other HIC columns on the market.
Unique Chemistry Designed for Optimal Selectivity of mAbs
The MAbPac HIC-10 column is based on high-purity, spherical, wide-pore (1,000Å), 5μm silica particles functionalized with proprietary alkyl amide groups. Its advanced surface bonding technology leads to excellent chemical stability, high efficiency, and unique selectivity optimized for mAbs and different from other HIC phases.- Excellent biocompatibility and extremely low carryover
- High resolution and high efficiency
- Compatible with both organic solvent and aqueous mobile phases
- Rugged column stability
Specifications
Analytical Column | |
250 mm | |
5 μm | |
1000 Å | |
Silica | |
Spherical, Fully Porous, Ultrapure Silica | |
Hydrophobic Interaction |
4.6 mm | |
8000 psi (550 bar) | |
2 to 8 | |
60°C | |
MAbPac | |
Hydrophobic Interaction | |
Bio-LC |