Learn More
MAbPac™ Capillary Reversed Phase HPLC Column
Description
Use this capillary column for high sensitivity HPLC or LC/MS characterization of intact proteins in top-down proteomics where sample amount is critically limited.
Use the Thermo Scientific™ MAbPac™ Capillary Reversed Phase HPLC Column high sensitivity HPLC or LC/MS characterization of intact proteins in top-down proteomics applications where sample amount is critically limited. This versatile column can be used for peptide-mapping characterization where the capillary format balances sensitivity with throughput. Additional applications are monoclonal antibodies (mAbs), fragments, variants, antibody drug conjugates (ADCs), PEGylated proteins, and bispecific proteins. These columns have excellent resolution, long column lifetime, and low carry-over.
Specifications
Specifications
| Column Format | Capillary HPLC Column |
| Diameter (Metric) | 0.15 mm |
| For Use With | High sensitivity HPLC or LC-MS of intact proteins in top-down proteomics with limited sample |
| Length (Metric) | 150 mm |
| Max. Pressure | 8700 psi (600 bar) |
| Particle Shape | N/A |
| Particle Size | 4 μm |
| pH | 0 to 9 |
| Pore Size | 1500 Å |
| Surface Area | N/A |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.