missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MabCaptureC Affinity Matrix

Product Code. 30274933 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
250 mL
1 L
5 L
10 L
Unit Size:
1 L
10 L
250mL
5 L
Product Code. Quantity unitSize
30274933 10 L 10 L
30019889 250 mL 250mL
30019838 1 L 1 L
30019782 5 L 5 L
4 options
This item is not returnable. View return policy

Product Code. 30274933

Brand: Thermo Scientific™ 196366210L

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ MabCaptureC Affinity Matrix is an agarose-based, high-binding-capacity Protein A chromatography resin with a rProtein A ligand produced in yeast.

MabCaptureC Affinity Matrix is an agarose-based, high-binding-capacity Protein A chromatography resin with a rProtein A ligand produced in yeast. The engineered rProtein A binds at the CH2-CH3 interface of the Fc-region of IgGs and shows cross binding to the VH3 domain of IgGs.

MabCaptureC™ Affinity Matrix is suitable for the purification of monoclonal antibodies.The rProtein A ligand is immobilized on a highly cross-linked and uniform-sized agarose-based bead with an average particle size of 75 μm. The uniform particle size delivers superior performance characteristics compared to traditional Protein A resins.

Features of the MabCaptureC™ Affinity Matrix include:

  • High dynamic binding capacity for mAbs of >50 g/L at 4.8 min residence time
  • Purification of mAbs and a subpopulation of scFv's and Fab fragments containing VH3 binding domains
  • Clean and sanitize with standard sodium hydroxide agents for chromatography products
  • Excellent scalability
  • Non-animal derived

Main characteristics

  • Matrix: agarose-based, epoxide activated
  • Average particle size: 75 ± 10 μm
  • Ligand: recombinant C domain of Protein A
  • Ligand coupling method: epoxide coupling
  • Binding capacity: >50 g IgG/L resin at 4.8 min residence time and 10% breakthrough
  • Elution conditions: 0.1 M glycine pH 3.0, or 20–100 mM sodium citrate or acetate, pH 3.0
  • Cleaning conditions: 0.2 M sodium hydroxide for 15 minutes every cycle
  • Operating pressure: ≤3 bar
  • Pressure flow: >500 cm/h at 3 bar in a 2.6 x 20 cm column
  • Formulation buffer: 20% (v/v) ethanol

For process development and resin screening, MabCaptureC™ is also available in the following formats:

  • MabCaptureC™ MiniChrom Column, 1 mL (Cat. No. 5943662001)
  • MabCaptureC™ MiniChrom Column for process development, 5 mL (5943662005)
  • MabCaptureC™ RoboColumn, 200 μL (5943662200)
  • MabCaptureC™ RoboColumn, 600 μL (5943662600)

Specifications

Format Container
Binding Capacity >50 g IgG/L
Ligand Engineered rProtein A
Matrix Agarose-based, epoxide-activated
Particle Size 75 ±10 μm
Target CH2-CH3 interface Fc-region
For Use With (Application) Monoclonal antibody purification
Quantity 10 L
Shipping Condition Ships at 2°C to 8°C
Form Agarose Bead

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.