missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAB21L3 Antibody (OTI2A3), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72562AF488
This item is not returnable.
View return policy
Description
MAB21L3 Monoclonal specifically detects MAB21L3 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| MAB21L3 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MAB21L3 | |
| Full length recombinant protein of human MAB21L3 (NP_689580) produced in HEK293T cells. | |
| 0.1 mL | |
| 126868.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI2A3 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| C1orf161, FLJ38716, mab-21-like 3 (C. elegans), protein mab-21-like 3, RP5-1185H19.1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction