missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAB21L2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10971-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is supposed to recognize both MAB21L2 and its homolog MAB21L1.
Required for eye morphogenesis. May promote the survival of proliferating retinal progenitor cells.Specifications
| MAB21L2 | |
| Polyclonal | |
| Unconjugated | |
| MAB21L2 | |
| mab 21 like 2 (C. elegans), MAB21L2, Protein mab 21 like 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10586, 23937, 680102 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8BPP1, Q9Y586 | |
| MAB21L2 | |
| MAB21L2 Fusion Protein Ag1423 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction