missing translation for 'onlineSavingsMsg'
Learn More

MAATS1 Antibody (OTI6G1), DyLight 594, Novus Biologicals™

Product Code. 18426677 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18426677 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18426677 Supplier Novus Biologicals Supplier No. NBP272309DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MAATS1 Monoclonal antibody specifically detects MAATS1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAATS1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI6G1
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias AAT1, AAT-1, AAT1alpha, AAT1-alpha, AAT1DKFZp781A2221, AMY-1-associating protein expressed in testis 1, C3orf15, chromosome 3 open reading frame 15, MYCBP/AMY-1-associated, testis expressed 1, MYCBP-binding protein, SPATA26
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-313 of human C3orf15 (NP_203528) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89876
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.