Learn More
Tocris Bioscience™ MA 2029
Potent and selective, competitive motilin receptor antagonist; orally active
Specifications
Purity >98%
View More Specs
Description
Description
Potent and selective, competitive motilin receptor antagonist (IC50 = 4.9 nM). Selective for the motilin receptor over a range of other receptors and ion channels. Inhibits motilin-induced duodenal muscle contractions in vitro. Also inhibits motilin-induced colonic and abdominal contractions in vivo. Orally active.
Specifications
Specifications
>98%