missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ M9 Minimal Medium Salts

Product Code. 11387689 Shop All MP Biomedicals Products
Change view
Click to view available options
:
454g(1lb)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11387689 454g(1lb)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11387689 Supplier MP Biomedicals™ Supplier No. 113037022

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

M9 Minimal Medium Salts (Maniatis Formulation) (Powder)

  • M9 Minimal Medium Salts can be made as a sterile 5X concentrated solution and diluted to 1X before use.
  • M9 Minimal medium salts are suitable for the preparation of M9 Minimal Medium for use in propagation of E. coli and plasmid amplification.
  • The resulting M9 medium can be used for maintaining positive selection pressure on plasmids coding for genes to produce essential substances such as amino acids and vitamins and for maintenance of F'-containing bacteria strains for M13 propagation.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Content And Storage Store M9 Minimal Medium Salts at ambient temperature (15-30°C). Medium is hygroscopic, therefore minimize exposure to air to prolong storage of media. Store sterile medium at 4°C.
Form Powder
Product Type Microbiology Media
Sterility Non-sterile
Formulation 33.9g Na2HPO4, 15g KH2PO4, 2.5g NaCl, 5g NH4Cl = 56.4g/L of a 5x solution.
For Use With (Application) Growth of E. coli cells
Quantity 454 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.