missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ M802 Humanized Recombinant Human Monoclonal Antibody (M802)
Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA554935
This item is not returnable.
View return policy
Description
Compatible with Direct ELISA.
M802 is an antibody-drug conjugate (ADC) that targets the HER2 receptor on cancer cells, delivering a cytotoxic agent to induce cell death.
Specifications
| M802 Humanized | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Human | |
| Protein A/G | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA | |
| M802 | |
| Unconjugated | |
| M-802; YZY Biopharma | |
| Recombinant Human CD340/ERBB2/HER2 and CD3E. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction