missing translation for 'onlineSavingsMsg'
Learn More
Learn More
m6A Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
£119.00 - £295.00
Specifications
| Antigen | m6A |
|---|---|
| Clone | 1D5E10 |
| Concentration | 1000 μg/mL |
| Applications | RNA Immunoprecipitation, Immunohistochemistry, ELISA, Dot Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30235385
|
Proteintech
68055-1-IG-20UL |
20 μL |
£135.00 £119.00 / 20µL Save £16.00 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30236320
|
Proteintech
68055-1-IG-150UL |
150 μL |
£349.00 £295.00 / 150µL Save £54.00 15% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The m6A antibody from Proteintech is a mouse monoclonal antibody generated with compound of m6A. This antibody recognizes m6a, human, mouse, rat antigen. The m6A antibody has been validated for the following applications: RIP, IHC, ELISA, Dot Blot analysis.Specifications
| m6A | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| Compound | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 1D5E10 | |
| RNA Immunoprecipitation, Immunohistochemistry, ELISA, Dot Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| IgG3 | |
| Protein A purification | |
| Monoclonal Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title