missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Single nanoViper™ and Double nanoViper™ PepMap™ Neo UHPLC Columns

Product Code. 17467583 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
150 mm
500 mm
750 mm
Product Line:
Double nanoViper
Single nanoViper
Unit Size:
1 unit
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Product Line unitSize
17467583 750 mm Double nanoViper Each
17447583 150 mm Double nanoViper Each
17457583 500 mm Double nanoViper Each
30040296 150 mm Single nanoViper 1 unit
30040297 150 mm Double nanoViper 1 unit
30040298 750 mm Single nanoViper 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 17467583 Supplier Thermo Scientific™ Supplier No. DNV75750PN
 Restricted Item
Estimated Shipment: 21-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Separate challenging peptide mapping samples with Thermo Scientific™ Double nanoViper™ PepMap™ Neo UHPLC columns.

Separate challenging peptide mapping samples with Thermo Scientific™ Single nanoViper™ and Double nanoViper™ PepMap™ Neo UHPLC columns. These columns feature flexible connectivity, high reproducibilty, and excellent separation performance. Our PepMap Neo columns are packed to higher pressure and provide 1500 bar pressure capability, improved column-to-column consistency, and increased efficiency. The column media is manufactured and selected to exacting standards and packed at high pressure, resulting in enhanced peak symmetry, resolution, and column-to-column reproducibility that allows you to reliably obtain greater sample coverage.

Benefits of PepMap Neo Columns

  • Brilliant Connectivity — Double nanoViper™ columns connect with ease.
  • Brilliant Reproducibility — Precise manufacturing protocols for packing media improve column-to-column consistency for reproducible analysis every day.
  • Brilliant Separation — Ultra-high pressure packing increases column efficiency, allowing you to run at up to 1500 bar for enhanced peak symmetry and resolution, providing higher sample capacity for more sample insights.
TRUSTED_SUSTAINABILITY

Specifications

Column Format Analytical Column, Double nanoViper™
Diameter (Metric) 75 μm
Length (Metric) 750 mm
Max. Pressure 22,000 psi (1500 bar)
Particle Size 2 μm
pH 2 to 8
Pore Size 100 Å
Quantity Each
Temperature 60°C
USP Type L1
Stationary Phase C18
Product Line Double nanoViper
Packing Material Spherical, Fully Porous, Ultrapure Silica
Carbon Load 15%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Description Double nanoViper™ PepMap™ Neo 2 μm C18 75 μm X 750 mm
For Use With (Application) Peptide Mapping, LC-MS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.