missing translation for 'onlineSavingsMsg'
Learn More

M3258, MedChemExpress

Product Code. 16484528
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16484528 10 mg 10mg
16417247 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16484528 Supplier Medchem Express Supplier No. HY111790
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress M3258 is an orally bioavailable, potent, reversible and highly selective immunoproteasome subunit LMP7 (β5i) inhibitor.

M3258 exerts high biochemical (IC50=3.6 nM) and cellular (IC50=3.4 nM) potency against the LMP7 subunit. M3258 shows strong antitumor efficacy in multiple myeloma xenograft models. M3258 leads to a significant and prolonged suppression of tumor LMP7 activity and ubiquitinated protein turnover and the induction of apoptosis in multiple myeloma cells.

Specifications

Chemical Name or Material M3258
CAS 2285330-15-4
Color White
Purity Grade Notes Research
Molecular Formula C17H20BNO5
Quantity 10 mg
Solubility Information DMSO : 250 mg/mL (759.51 mM; Need ultrasonic)
SMILES O=C(N[C@@H](CC1=COC2=C1C=CC=C2)B(O)O)[C@H]3[C@@H]4CC[C@@H](O4)C3
Molecular Weight (g/mol) 329.16
Formula Weight 329.16
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.