missing translation for 'onlineSavingsMsg'
Learn More

Cytiva M13KO7 Helper Phage

Product Code. 10439944 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10439944 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10439944 Supplier Cytiva Supplier No. 27152401

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For phage rescue

  • Contains a modified gene 11, an insertion of the origin of replication of p15A, and the kanamycin resistance gene from Tn903 at the Ava I site
  • Supplied ready to use at >5 × 1010 pfu/mL

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Quantity 100 μL
For Use With (Application) Phage rescue
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.