missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ M13 Phage coat protein Monoclonal Antibody (4B1D7)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA550272
This item is not returnable.
View return policy
Description
M13 Phage coat protein Monoclonal Antibody for Western Blot, ELISA
M13 antibodies have numerous uses in screening phage display libraries, especially the screening of single chain antibody libraries. The display of repertoires of antibody fragments on the surface of filamentous phage offers a new way to produce immunoreagents with defined specificities. Phage derived antibody fragments offer a number of advantages over mouse monoclonal antibodies, such as better clearance from the blood, the possibility to select from human combinatorial libraries and the relative ease by which such fragments can be manipulated. The phage display technique thus facilitates the selection of antibody fragments of therapeutic value or research interest. Antibodies to M13 filamentous phage coat proteins are instrumental in the selection and detection of phages expressing specific antibody fragments or peptide sequences at their surface.
Specifications
| M13 Phage coat protein | |
| Monoclonal | |
| 2 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| Mouse | |
| Protein G | |
| RUO | |
| Virus | |
| Antibody | |
| IgG1 |
| ELISA, Western Blot | |
| 4B1D7 | |
| Unconjugated | |
| Attachment protein G3P; G3P; Gene 3 protein; III; Minor coat protein | |
| Recombinant Enterobacteria phage M13 Attachment protein (19-424AA). | |
| 100 μL | |
| Primary | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction