missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ M13 Human, Reverse Primer, Desalted

Product Code. 15524374 Shop All Applied Biosystems Products
Click to view available options
Quantity:
25 nmol
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15524374

Brand: Applied Biosystems™ A15634

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

M13 Human, Reverse Primer, Desalted

Use our tool to find and order pre-designed primers for PCR and Sanger sequencing. Free of SNPs and dimers, target-specific, and suitable for universal PCR conditions, these primers can be ordered unmodified, M13-tailed, HPLC-purified, or desalted. All primers are checked by mass spectrometry.
TRUSTED_SUSTAINABILITY

Specifications

Format Solid
Description M13 Human Reverse Desalted Primer, 25 nmole, Solid, For PCR and Sanger Sequencing
Product Type M13 Human Reverse DS Primer
Purification Method Desalted
Purity Desalted
Primer Reverse
Quantity 25 nmol
Grade HPLC
Form Solid

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.