missing translation for 'onlineSavingsMsg'
Learn More

M13 bacteriophage Antibody (E1), DyLight 550, Novus Biologicals™

Product Code. 18430267 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18430267 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18430267 Supplier Novus Biologicals Supplier No. NBP266601R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

M13 bacteriophage Monoclonal antibody specifically detects M13 bacteriophage in Virus samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen M13 bacteriophage
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone E1
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias Coat protein A; Coat protein B; Coat protein C polypeptide I; Coat protein C polypeptide II; Coat protein D; G3P; III; IX; M13 coat protein A; M13 coat protein B; M13 coat protein C polypeptide I; M13 coat protein C polypeptide II; M13 coat protein D; M13
Host Species Mouse
Immunogen Hybridoma produced by the fusion of splenocytes from mice immunized with isolated M13 phage coat proteins and mouse myeloma cells.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.