missing translation for 'onlineSavingsMsg'
Learn More

m-Xylylene dichloride, 97%

Product Code. 11414583
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11414583 100 g 100g
11418790 10 g 10g
11404583 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11414583 Supplier Thermo Scientific Alfa Aesar Supplier No. A11548.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 626-16-4 | C8H8Cl2 | 175.052 g/mol

m-Xylylene dichloride is used in pharmaceutical and used in medicine, brightener intermediates. α,α?-Dichloro-m-xylene is suitable for use in the synthesis of 1,7-bis(6-methoxy-2-formylphenyl)-1,7-dioxaheptane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
m-Xylylene dichloride is used in pharmaceutical and used in medicine, brightener intermediates. α,α′-Dichloro-m-xylene is suitable for use in the synthesis of 1,7-bis(6-methoxy-2-formylphenyl)-1,7-dioxaheptane.

Solubility
Insoluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-16-4
Molecular Formula C8H8Cl2
Molecular Weight (g/mol) 175.052
MDL Number MFCD00000912
InChI Key GRJWOKACBGZOKT-UHFFFAOYSA-N
Synonym 1,3-bis chloromethyl benzene, alpha,alpha'-dichloro-m-xylene, m-xylylene dichloride, benzene, 1,3-bis chloromethyl, m-bis chloromethyl benzene, m-xylylene chloride, m-dichloroxylene, benzene, 1,3-dichloromethyl, ccris 1774, m-xylene, .alpha.,.alpha.'-dichloro
PubChem CID 12275
IUPAC Name 1,3-bis(chloromethyl)benzene
SMILES C1=CC(=CC(=C1)CCl)CCl

Specifications

Melting Point 33°C to 37°C
Density 1.302
Boiling Point 250°C to 255°C
Flash Point 144°C (291°F)
Quantity 100 g
UN Number UN2928
Beilstein 2079898
Solubility Information Insoluble in water.
Formula Weight 175.06
Percent Purity 97%
Chemical Name or Material m-Xylylene dichloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.