missing translation for 'onlineSavingsMsg'
Learn More

m-Xylene, TRC

Product Code. 30451264
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30451264 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 30451264 Supplier Toronto Research Chemicals Supplier No. TRCX749415250ML

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material m-Xylene (1,3-Dimethylbenzene)
CAS 108-38-3
Molecular Formula C8 H10
Quantity 250 mL
Synonym Benzene','1,3-dimethyl-, m-Xylene (8CI), 1,3-Dimethylbenzene, 1,3-Xylene, 3-Methyltoluene, NSC 61769, m-Dimethylbenzene, m-Methyltoluene, m-xylol
InChI Formula InChI=1S/C8H10/c1-7-4-3-5-8(2)6-7/h3-6H,1-2H3
SMILES Cc1cccc(C)c1
IUPAC Name 1,3-xylene
Molecular Weight (g/mol) 106.16
Formula Weight 106.0783
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.