missing translation for 'onlineSavingsMsg'
Learn More

m-Tolylhydrazine hydrochloride, 98%

Product Code. 11480264
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11480264 5 g 5g
11490264 25 g 25g
11400274 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11480264 Supplier Thermo Scientific Alfa Aesar Supplier No. A13667.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 637-04-7 | C7H11ClN2 | 158.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 637-04-7
Molecular Formula C7H11ClN2
Molecular Weight (g/mol) 158.63
MDL Number MFCD00012932
InChI Key SHTBTGXNVWYODU-UHFFFAOYSA-N
Synonym 3-methylphenylhydrazine hydrochloride, m-tolylhydrazine hydrochloride, m-tolylhydrazinium 1+ chloride, 3-methylphenyl hydrazine hydrochloride, 3-methylphenylhydrazine hcl, m-tolyhydrazine hydrochloride, 1-3-methylphenyl hydrazine hydrochloride, 3-tolylhydrazine hydrochloride, 3-methyl phenylhydrazine hydrochloride, hydrazine, 3-methylphenyl-, monohydrochloride
PubChem CID 71562
IUPAC Name (3-methylphenyl)hydrazine;hydrochloride
SMILES [H+].[Cl-].CC1=CC=CC(NN)=C1

Specifications

Melting Point ∼193°C (decomposition)
Quantity 5 g
UN Number UN2811
Beilstein 3563995
Formula Weight 158.63
Percent Purity 98%
Chemical Name or Material m-Tolylhydrazine hydrochloride

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.