missing translation for 'onlineSavingsMsg'
Learn More

m-Tolylacetyl chloride, 99%, Thermo Scientific™

£57.76 - £162.00

Chemical Identifiers

CAS 13910-79-7
Molecular Formula C9H9ClO
Molecular Weight (g/mol) 168.62
MDL Number MFCD03424715
InChI Key BGGKEDDFKBVTDK-UHFFFAOYSA-N
Synonym 2-3-methylphenyl acetyl chloride, m-tolylacetyl chloride, m-tolyl-acetyl chloride, 3-methylphenyl acetyl chloride, benzeneacetyl chloride,3-methyl, 3-methylbenzeneacetyl chloride
PubChem CID 2760633
IUPAC Name 2-(3-methylphenyl)acetyl chloride
SMILES CC1=CC(=CC=C1)CC(=O)Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15467777
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H37453.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15457777
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H37453.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

13910-79-7
168.62
BGGKEDDFKBVTDK-UHFFFAOYSA-N
2760633
CC1=CC(=CC=C1)CC(=O)Cl
C9H9ClO
MFCD03424715
2-3-methylphenyl acetyl chloride, m-tolylacetyl chloride, m-tolyl-acetyl chloride, 3-methylphenyl acetyl chloride, benzeneacetyl chloride,3-methyl, 3-methylbenzeneacetyl chloride
2-(3-methylphenyl)acetyl chloride

Specifications

13910-79-7
98°C (7.5 mmHg)
MFCD03424715
Moisture sensitive
BGGKEDDFKBVTDK-UHFFFAOYSA-N
2-(3-methylphenyl)acetyl chloride
2760633
99%
m-Tolylacetyl chloride
Red
C9H9ClO
UN3265
2-3-methylphenyl acetyl chloride, m-tolylacetyl chloride, m-tolyl-acetyl chloride, 3-methylphenyl acetyl chloride, benzeneacetyl chloride,3-methyl, 3-methylbenzeneacetyl chloride
CC1=CC(=CC=C1)CC(=O)Cl
168.62
168.62
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications