missing translation for 'onlineSavingsMsg'
Learn More

m-Tolylacetic acid, TRC

Product Code. 30451350
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30451350 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30451350 Supplier Toronto Research Chemicals Supplier No. TRCT5365801G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material m-Tolylacetic acid
CAS 621-36-3
Molecular Formula C9H10O2
Quantity 1 g
Synonym 3-Methylbenzeneacetic acid, (3-Methylphenyl)acetic acid, 2-(3-Methylphenyl)acetic acid, 2-(m-Tolyl)acetic acid, 3-Tolylacetic acid, m-Methylbenzeneacetic acid, m-Methylphenylacetic acid
InChI Formula InChI=1S/C9H10O2/c1-7-3-2-4-8(5-7)6-9(10)11/h2-5H,6H2,1H3,(H,10,11)
SMILES Cc1cccc(CC(=O)O)c1
IUPAC Name 2-(3-methylphenyl)acetic acid
Molecular Weight (g/mol) 150.17
Formula Weight 150.0681
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.