missing translation for 'onlineSavingsMsg'
Learn More
Learn More
m-Toluoyl Chloride, TRC
Specifications
Specifications
| Chemical Name or Material | m-Toluoyl Chloride |
| CAS | 1711-06-4 |
| Molecular Formula | C8H7ClO |
| Quantity | 5 g |
| Synonym | 3-Methylbenzoyl Chloride, 3-Toluoyl Chloride, NSC 97207, Toluoyl Chloride, m-Methylbenzoyl Chloride, m-Toluenecarbonyl Chloride, m-Toluic Acid Chloride, m-Toluyl Chloride |
| InChI Formula | InChI=1S/C8H7ClO/c1-6-3-2-4-7(5-6)8(9)10/h2-5H,1H3 |
| SMILES | Cc1cccc(c1)C(=O)Cl |
| IUPAC Name | 3-methylbenzoyl chloride |
| Molecular Weight (g/mol) | 154.59 |
| Formula Weight | 154.0185 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?